missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, Biological Stain, C.I.52015, Fisher Chemical

Brand:  Fisher Chemical M/P206/46


Product Code. 10539000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

25g, Glass Jar

TRUSTED_SUSTAINABILITY
Specifications

Specifications

190°C
C16H18ClN3S
CXKWCBBOMKCUKX-UHFFFAOYSA-M
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
6099
319.85g/mol
Green
25 g
61-73-4
12111
CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]
319.851
CHEBI:6872
Glass Jar
3 to 4.5
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methylene Blue

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers