missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylene Blue hydrate, 96+%, high purity biological stain
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Melting Point | 100.0°C to 110.0°C |
Dye Content | 96.0 to 103.0% (on dried substance) |
Assay Percent Range | 96+% |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol |
Infrared Spectrum | Authentic |
Formula Weight | 319.85 |
Loss on Drying | 8.0% to 22.0% (105°C) |
Percent Purity | ≥96% |
Packaging | Glass bottle |
Color | Green |
Show More |
Safety and Handling
Product Identifier
- Methylene Blue hydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- MIXTURE LIST-Contains : Methylene Blue hydrate, Methylene blue
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction