missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylene Blue, pure, certified
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 190.0°C |
Absorption Ratio | 1.21 to 1.7 |
Dye Content | 82% min. |
Beilstein | 27, V,19, 259 |
Merck Index | 15, 6132 |
Solubility Information | Solubility in water: 40g/L (20°C). Other solubilities: in methanol: 430g/L |
Infrared Spectrum | Authentic |
Lambda Max | 663 to 667nm |
Formula Weight | 319.85 |
Grade | Pure |
Show More |
Safety and Handling
Product Identifier
- Methylene Blue
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction