missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, Redox Indicator, Solid, C.I.52015, Fisher Chemical™

Product Code. 10010270
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
PubChem CID 104827
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point 100°C
Formula Weight 373.89g/mol
Packaging Glass Jar
Color Green
pH 3 to 4.5
Quantity 10 g
Physical Form Solid
compliance-icons
Product Identifier
  • Methylene Blue trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Methylene blue trihydrate, Methylene blue
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'