missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylenediphosphonic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Methylenediphosphonic acid is used as a radioactive imaging agent, which is primarily used in skeletal scintigraphy. Due to its absorption by a variety of tumors, it is useful for the detection of neoplasms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methylenediphosphonic acid is used as a radioactive imaging agent, which is primarily used in skeletal scintigraphy. Due to its absorption by a variety of tumors, it is useful for the detection of neoplasms.
Solubility
Soluble in water, dimethylsulfoxide and methanol. Slightly soluble in anhydrous ethanol. Insoluble in methylene chloride.
Notes
Store in a cool place. Incompatible with strong bases.
Methylenediphosphonic acid is used as a radioactive imaging agent, which is primarily used in skeletal scintigraphy. Due to its absorption by a variety of tumors, it is useful for the detection of neoplasms.
Solubility
Soluble in water, dimethylsulfoxide and methanol. Slightly soluble in anhydrous ethanol. Insoluble in methylene chloride.
Notes
Store in a cool place. Incompatible with strong bases.
Specifications
Specifications
Melting Point | 202°C to 204°C |
Odor | Odorless |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 1708494 |
Sensitivity | Hygroscopic |
Merck Index | 14,5792 |
Solubility Information | Soluble in water,dimethylsulfoxide and methanol. Slightly soluble in anhydrous ethanol. Insoluble in methylene chloride. |
Formula Weight | 176 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
Product Identifier
- Methylenediphosphonic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Medronic acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'