missing translation for 'onlineSavingsMsg'
Learn More

Methylethiocyanate, 99%, ACROS Organics™

Product Code. 10357580
Click to view available options
5mL; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 556-64-9
Color Colorless to Yellow
Density 1.068g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C2H3NS
Linear Formula CH3SCN
Refractive Index 1.4670 to 1.4690 (20°C, 589nm)
MDL Number MFCD00001830
Quantity 5 mL
Beilstein 03, 175
Merck Index 15, 6200
Synonym methyl sulfocyanate, methylrhodanid, thiocyanic acid, methyl ester, methyl rhodanate, methylthiokyanat, methane, thiocyanato, thiocyanatomethane, methylthiocyanate, thiocyanic acid methyl ester, ch3scn
InChI Key VYHVQEYOFIYNJP-UHFFFAOYSA-N
SMILES CSC#N
IUPAC Name methyl thiocyanate
Molecular Weight (g/mol) 73.113
PubChem CID 11168
ChEBI CHEBI:61112
Formula Weight 73.11
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Methylethiocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.