Learn More
Methylmagnesium bromide, 1M to 1.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Brown-Gray to Brown-Purple |
Linear Formula | CH3MgBr |
Concentration | 0.95 to 1.20M |
Density | 1.0000g/mL |
Solubility Information | Solubility in water: vigorous reaction. |
Formula Weight | 119.24 |
Packaging | AcroSeal™ Glass Bottle |
Specific Gravity | 1 |
Quantity | 800 mL |
Flash Point | −21°C |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0440332 | Certificate of Analysis | 31/01/2025 | 377381000,377388000 |
A0440332 | Certificate of Analysis | 28/11/2023 | 377381000,377388000 |
A0399191 | Certificate of Analysis | 14/12/2022 | 377381000,377388000 |
A0394395 | Certificate of Analysis | 14/12/2022 | 377381000,377388000 |
A0430977 | Certificate of Analysis | 19/01/2022 | 377381000,377388000 |
Safety and Handling




- Methylmagnesium bromide, 1M solution in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.