missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™

Product Code. 10434862 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10434862

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-16-1, 60-29-7
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent
PubChem CID 6349
SMILES C[Mg]Br

Specifications

Color Colorless to Yellow
Linear Formula CH3MgBr
Concentration 2.85 to 3.20M
Density 1.0350g/mL
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name magnesium;carbanide;bromide
Formula Weight 119.26
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.035
Quantity 100 mL
Flash Point −40°C
Physical Form Solution, may develop some turbidity precipitate
Chemical Name or Material Methylmagnesium bromide, 3M Solution in Diethyl Ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylmagnesium bromide, 3M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • EUH019-May form explosive peroxides.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P223-Do not allow contact with water.
Supplemental information
  • MIXTURE LIST-Contains : Methyl magnesium bromide, Ethyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.