missing translation for 'onlineSavingsMsg'
Learn More
Methylmagnesium chloride, 3M (22 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10560522

Methylmagnesium chloride, 3M (22 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10560522
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10560522

Brand: Thermo Scientific Acros 252561000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-58-4, 109-99-9
Molecular Formula CH3ClMg
Molecular Weight (g/mol) 74.79
MDL Number MFCD00000468
InChI Key RQNMYNYHBQQZSP-UHFFFAOYSA-M
Synonym methylmagnesium chloride, chloromethylmagnesium, chloro methyl magnesium, magnesium, chloromethyl, memgcl, methylmagnesiumchloride, methyl magnesium chloride, unii-m5e1132g4w, ch3mgcl, qmabhcaih@
PubChem CID 12670
IUPAC Name chloro(methyl)magnesium
SMILES C[Mg]Cl

Specifications

Color Brown
Boiling Point 66.0°C
Linear Formula CH3MgCl
Density 1.0100g/mL
Solubility Information Solubility in water: reacts.
Formula Weight 74.79
Percent Purity 22 to 24%
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.01
Quantity 100 mL
Flash Point −17°C
Physical Form Liquid
Chemical Name or Material Methylmagnesium chloride, 22 wt. % Solution in Tetrahydrofuran
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465810 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number252561000,252568000
Lot NumberA0453877 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number252561000,252568000
Lot NumberA0451420 Certificate TypeCertificate of Analysis Date02/10/2023 Catalog Number252561000,252568000
Lot NumberA0448965 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog Number252561000,252568000
Lot NumberA0397712 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number252561000,252568000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylmagnesium chloride, 22 wt% solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methylmagnesium chloride, 3M (22 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.