missing translation for 'onlineSavingsMsg'
Learn More

Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals

Product Code. 15451037
Click to view available options
Quantity:
1 mL
10 mL
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

As a defoliant and epoxidation catalyst, as denaturing agent

Methyl mercury(II) hydroxide is used as defoliant and epoxidation catalyst, as denaturing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-57-2
MDL Number MFCD00013924

Specifications

Color Colorless
Physical Form Liquid
Quantity 10 mL
Linear Formula CH3HgOH
UN Number UN2024
Solubility Information Fully miscible in water.
Formula Weight 232.63
Concentration or Composition (by Analyte or Components) 1M in H2O
Chemical Name or Material Methylmercury(II) hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.