missing translation for 'onlineSavingsMsg'
Learn More

Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals

Product Code. 15461037
Click to view available options
Quantity:
1 mL
10 mL
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

As a defoliant and epoxidation catalyst, as denaturing agent

Methyl mercury(II) hydroxide is used as defoliant and epoxidation catalyst, as denaturing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-57-2
MDL Number MFCD00013924

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Linear Formula CH3HgOH
UN Number UN2024
Solubility Information Fully miscible in water.
Formula Weight 232.63
Concentration or Composition (by Analyte or Components) 1M in H2O
Chemical Name or Material Methylmercury(II) hydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'