missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylthymol blue, sodium salt , indicator grade

Product Code. 10560744
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1945-77-3
Molecular Formula C37H44N2Na4O13S
Molecular Weight (g/mol) 848.779
MDL Number MFCD00148905
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
PubChem CID 131850582
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Specifications

Solubility Information Solubility in water: 460g/L (20°C).
Infrared Spectrum Authentic
Formula Weight 844.76
Grade ACS Reagent
Packaging Glass bottle
Suitability pH 6.5 (yellow) to pH 8.5 (blue):Pass Test
Color Red
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Methylthymol blue, tetrasodium salt
Product Identifier
  • Methylthymol Blue, tetrasodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'