missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyltin trichloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A chemical used in the synthesis of metallasilesesquiloxanes. Also used in the preparation of organotin(IV) complexes showing antibacterial activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 43°C |
Quantity | 5 g |
UN Number | UN2923 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in Chloroform,Methanol. |
Formula Weight | 240.11 |
Percent Purity | 97% |
Physical Form | Crystalline |
Chemical Name or Material | Methyltin trichloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu