missing translation for 'onlineSavingsMsg'
Learn More

Methyltrioxorhenium(VII), 98%

Product Code. p-7047270
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11345005

Brand: Thermo Scientific Alfa Aesar L18478.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 70197-13-6 | CH3O3Re- | 249.239 g/mol

Methyltrioxorhenium(VII) is used as a catalyst for various transformations such as olefin metathesis. It is associated with hydrogen peroxide and used as an oxidizing agent. It is used to prepare acid or ester from the corresponding terminalalkynes. It is a catalytic oxidant used with urea hydrogen peroxide to prepare nitrones. It reacts with 2-methylnaphthalene to get 2-methyl-1,4-naphthoquinone (Vitamin K3). Further, it catalyzes the reaction with aldehydes and diazoketones to get an alkene. In addition to this, it is used as an effective antioxidant under various conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltrioxorhenium(VII) is used as a catalyst for various transformations such as olefin metathesis. It is associated with hydrogen peroxide and used as an oxidizing agent. It is used to prepare acid or ester from the corresponding terminalalkynes. It is a catalytic oxidant used with urea hydrogen peroxide to prepare nitrones. It reacts with 2-methylnaphthalene to get 2-methyl-1,4-naphthoquinone (Vitamin K3). Further, it catalyzes the reaction with aldehydes and diazoketones to get an alkene. In addition to this, it is used as an effective antioxidant under various conditions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70197-13-6
Molecular Formula CH3O3Re-
Molecular Weight (g/mol) 249.239
MDL Number MFCD00192332
InChI Key VZSXFJPZOCRDPW-UHFFFAOYSA-N
Synonym methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii
PubChem CID 2734010
IUPAC Name carbanide;trioxorhenium
SMILES [CH3-].O=[Re](=O)=O

Specifications

Melting Point 110°C to 115°C
Quantity 100 mg
Linear Formula CH3ReO3
UN Number UN1479
Beilstein 3927932
Solubility Information Soluble in water.
Formula Weight 249.24
Percent Purity 98%
Sensitivity Light and moisture sensitive
Chemical Name or Material Methyltrioxorhenium(VII)

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado