missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11343126
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 791-29-7
Molecular Formula C19H18Si
Molecular Weight (g/mol) 274.438
MDL Number MFCD00041325
InChI Key GIGVICQLYWGMGW-UHFFFAOYSA-N
Synonym silane, methyltriphenyl, triphenylmethylsilane, methyl triphenyl silane, methyl-tri phenyl silane, methyltriphenylsilane
PubChem CID 522714
IUPAC Name methyl(triphenyl)silane
SMILES C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 66°C to 68°C
Quantity 1 g
Beilstein 2940360
Formula Weight 274.44
Percent Purity 97%
Chemical Name or Material Methyltriphenylsilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'