missing translation for 'onlineSavingsMsg'
Learn More

Metolazone, Fisher BioReagents

Product Code. 10728705
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Packaging:
Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy
Metolazone is an anti-hypertensive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17560-51-9
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00069304
InChI Key AQCHWTWZEMGIFD-UHFFFAOYNA-N
Synonym metolazone, zaroxolyn, diulo, mykrox, microx, metalozone, metenix, oldren, metolazona, metolazonum
PubChem CID 4170
ChEBI CHEBI:64354
SMILES CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O

Specifications

Melting Point 248°C
Color White
Packaging Amber Glass
Quantity 25 mg
Merck Index 15, 6224
IUPAC Name 7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2-dihydroquinazoline-6-sulfonamide
Formula Weight 365.8
Physical Form Powder or Solid
Chemical Name or Material Metolazone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'