Learn More
Emma Safety Footwear Metric XD Safety Shoes
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
Brand: Emma Safety Footwear MM50756047
This item is not returnable.
View return policy
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Easy Twist Outsole
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Metric XD | |
Microfiber | |
47 |