missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Metricel™ Black Membrane

Contrasting membrane for microbiological analysis. Dark background provides excellent contrast for counting opaque colonies in microbiology labs.

Brand:  Cytiva (formerly Pall Lab) 66585

298.30 EUR valid until 2024-12-31
Use promo code "21931" to get your promotional price.


 View more versions of this product

Product Code. 17124581

  • 314.00 € / Pack of 200
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Exclusive dot grid pattern does not enhance or inhibit colony growth certified for use in the Membrane Filter (MF) Technique
  • For research only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black
Metricel Black Membrane Modified PES
Sterile
Yes
Gridded
Bulk with Parchment Separators
0.45 μm
White
200/Pk.
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers