missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Cantilever Shelves

For use with Super Erecta™ or HD Super™ shelves

Brand:  Metro™ 1248CSNC

Product Code. 15966551

  • 72.40 € / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Drop mat design creates a retaining ledge around the entire deep shelf
  • For use with Super Erecta™ or HD Super™ shelves
  • Width: 12 in.

Order Info

Order rear posts that are taller than front posts to accommodate this type of shelf.

Specifications

Specifications

48 x 12 in. (121.9 x 30.48 cm)
48 in.
12 in.
Chrome
Metro™ Super Erecta™ shelving system
121.9 cm
30.48 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers