missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Double-Deep qwikTRAK™ Track Sets
Create tracks up to 21 ft.
Brand: Metro™ LBTS6NA
This item is not returnable.
View return policy
Description
- For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q and MetroMax i shelving systems
- Sets include tracks and hardware needed to assemble system up to 21 ft. (640cm)
- 17-823-756 (LBTS2.5NA) includes a joining kit to connect the track to other track sets
Specifications
Track Set | |
182.88 cm | |
Tracks and hardware needed to assemble double deep qwikTRAK™ system up to 21 ft. | |
4.1 kg |
72 in. | |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system | |
9 lb. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Metro™ Double-Deep qwikTRAK™ Track Sets > Track length: 6 ft. (182.8cm)
Spot an opportunity for improvement?Share a Content Correction