missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Flexline™ Sharps Container Cart Accessory

Sharps accessories attach to carts for easy accessibility while mobile.

Brand:  Metro™ FL251

 View more versions of this product

Product Code. 15966911

  • 256.00 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sharps cart accessory in lockable or non-locking styles
  • Lockable dimensions with glove box: 12.06 x 33.02 x 50.48 cm (4.75 x 13 x 19.875 in.)
  • Lockable dimensions without glove box: 12.06 x 33.02 x 28.89 cm (4.75 x 13 x 11.375 in.)
  • Nonlocking dimensions: 24.1 x 8.3 x 23.7cm (9.5 x 3.25 9.3125 in.)
Specifications

Specifications

Red
Sharps Container Only
Epoxy-coated Steel, Plastic
Not Autoclavable
Lifeline Carts
Sharps Container
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.