missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MetroMax i Basket Glassware Cart

Level 2 cart with built-in Microban antimicrobial product protection allows arrangement configuration, ensuring storage and movement won't disrupt your workday.

Specifications

Product Type Glassware Cart
Capacity (Metric) 362.8 kg
Color Taupe, Blue
For Use With (Application) Glassware Transportation
Length (English) 36 in.
View More Specs

Description

Description

  • Modular design offers ultimate flexibility.
  • Reinforced polymers provide superior strength (up to 800 lb. per shelf).
  • Corrosion-proof, suitable for damp and humid conditions.
  • Lifetime warranty against rust and corrosion.
  • Shelves wipe down easily and have a retaining edge that contains drips and spills.
  • Two-basket and four-basket capacity glassware solutions are offered in a spacesaving 18 x 36 in. (457 x 914mm) footprint.
  • Solid MetroMax bottom shelf provides spill retention and can be removed from shelf frame for easy cleaning.
  • Wide range of accessories available separately, including all-polymer casters for maximum corrosion resistance and premium performance.

Order Info

Casters not included (5PC Series Metro casters are recommended for superior lifetime performance).

Specifications

Specifications

Glassware Cart
Taupe, Blue
36 in.
800 lb.
Polymer
45.7 cm
362.8 kg
Glassware Transportation
91.4 cm
362.8 kg
18 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.