missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax i™ Mobile Desk Lab Cart
Designed to offer superior corrosion resistance
Brand: Metro™ MDSX3
935.90 EUR valid until 2024-11-01
Use promo code "21705" to get your promotional price.
This item is not returnable.
View return policy
Description
- Smooth rolling
- 5 in. (12.7cm) polyurethane casters come standard with each cart
- All shelves adjust in .5 in. (1.27cm) increments
Specifications
Mobile Desk | |
Ideal for instrumentation storage or can be combined with accessories to provide a functional lab work surface, superior corrosion resistance | |
101.6 cm | |
36 in. | |
18 in. |
36 x 18 x 40 in. (91.4 x 45.7 x 101.6 cm) | |
40 in. | |
4 Polyurethane brake casters (127 mm) | |
91.4 cm | |
45.7 cm |