missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax iQ™ Accessory, Keyboard Shelf
For use with MetroMax iQ shelves
Brand: Metro™ KBX
This item is not returnable.
View return policy
Description
- Epoxy-coated shelf provides high level of corrosion resistance
- Mounts to 18 in. or 24 in. wide shelves
- Minimum shelf length is 30 in.
Shelf load should not exceed 10 lbs. (45.8kg).
Specifications
Keyboard Shelf | |
18 and 24 in.Wide MetroMax i Shelves | |
127 cm | |
67.3 cm | |
45.8 kg | |
40.6 cm |
26.5 x 16 x 5 in. | |
5 in. | |
26.5 in. | |
10 lb. | |
16 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction