missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Cart with Solid and Wire Shelves

Utility cart is a smart addition to any workplace featuring Super Erecta™ shelves with plastic split sleeves, two one-piece handles, and casters with donut bumpers.

Brand:  Metro™ MW404

 View more versions of this product

Product Code. 15987171

  • 447.00 € / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel, chrome shelf and handle material.
  • Load capacity: 375 lb. (170.1 kg).
  • Fully interchangeable Super Erecta™ shelves.
  • Center shelf can be positioned at any height in 1 in. (2.5 cm) increments.
  • One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability.
  • 18 in. (46 cm) carts have 4 in. diameter (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. diameter (13 cm) casters.
  • Three tiers, 375 lbs (170 kg) weight capacity per cart.
  • Solid Shelves 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength.
  • Raised edges help contain spills and make cleanup easier.
  • Sturdy corner castings facilitate mounting.
  • Wire Shelves chrome-plated or stainless-steel shelves with trademarked truss design for added strength.
  • Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place.

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Specifications

Utility Cart
NSF
3-tier Model
Transporting Materials
38 in.
Super Erecta™ 3 Chrome-plated wire shelves with plastic split sleeves, 2 one-piece Chrome-plated handle, 4 in. dia. caster, 3.5 in. donut bumpers
91 cm
170 kg
3
26.8 kg
21 in.
170 kg
Silver
36 x 21 x 38 in.
Chrome
97 cm
36 in.
375 lb.
Stainless Steel
Stainless Steel, Chrome
59 lb.
53 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.