Learn More
Metro™ Stainless Lab Worktable, Gray Phenolic Top and 3-Sided Frame
Lab worktable helps keep workers safe with a phenolic top and corrosion-proof, stainless steel construction throughout the body.
Brand:  Metro™ LTSM48UPG
 View more versions of this product
Product Code. 15957141
  • 1450.00 € / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
Mobile Table
47.75 x 30 x 36 in.
36 in.
3-sided Frame, Adaptor Insert
121.2 cm
273 kg
46 kg
76.2 cm
Silver
Laboratory
91.44 cm
47.75 in.
600 lb.
101 lb.
30 in.
Stainless Steel