Learn More
Metro™ Stainless Lab Worktable, Gray Phenolic Top and Solid HD Shelf
Metro Stainless Lab Worktable features corrosion-proof performance and stainless steel construction throughout.
Brand:  Metro™ LTSM48PG
 View more versions of this product
Product Code. 15917141
  • 1450.00 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
Mobile Lab Worktable
Moderate to high chemical use and have excellent abrasion resistance and impact resistance
90.93 cm
47.75 in.
600 lb.
Gray Phenolic Top or Gray Phenolic Top with Backsplash
30 in.
Stainless Steel
47.75 x 30 x 35.8 in.
35.8 in.
Stainless Steel HD Shelf, Adapter Insert, 4 HD Posts
121.28 cm
273 kg
Metro 5 in. Stem Casters
76.2 cm