Learn More
Metro™ Stainless Laboratory Worktable Accessory, Wire Shelf
Expand your workspace in your lab. Metro Stainless Laboratory Worktable shelves come in a choice of stainless steel or epoxy-coated.
Brand:  Metro™ LT48WS
 View more versions of this product
Product Code. 15937041
  • 250.00 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Shelves mount on three-sided frames with use of clips
  • Load rated for a maximum of 150 lbs.
  • Also compatible with MetroMax i Lab Worktables
Specifications
Specifications
Wire Shelf
Metro™ MetroMax i™ and other Stainless Steel worktables
102.5 cm
68 kg
10 lb.
Stainless
40.3 in.
150 lb.
3-Sided Frame
4.5 kg