missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Starsys™ Cart Accessories
Parts for use with Starsys carts
Brand: Metro™ SXRS6
This item is not returnable.
View return policy
Description
- 6 in. FL drawer
- No drawer pull
Specifications
Drawer | |
Metro Starsys cart | |
13.63 cm | |
40 cm | |
16.25 in. |
15.75 x 16.25 x 5.37 in. (40 x 14 x 13.63 cm) | |
5.37 in. | |
15.75 in. | |
Polymer | |
14 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction