missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Starsys™ Overhead Storage Unit

Microban™ antimicrobial protection prevents growth of stains and odor-causing bacteria.

Brand:  Metro™ SXRSOH27P2S

 View more versions of this product

Product Code. 15927271

  • 1330.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Built on same widths as Starsys base units utilizing very similar construction
  • Insures that wall cabinets align with lower product configuration in application
  • All overhead cabinets include corrugated polymer inner panels in their assembly as well as clean-design back panel, molded with cosmetic side facing inside of cabinet, creating much cleaner appearance - especially when used without doors or in conjunction with clear door
  • Necessary wall mount rails/hardware are included with each wall cabinet
  • Microban™ antimicrobial protection inhibits growth of stain and odor-causing bacteria on Starsys, keeping surface areas cleaner between cleanings
Specifications

Specifications

13.8 x 21.5 x 29.5 in.
29.5 in.
None
Taupe
29.5 in.
Overhead Storage Unit
Single
1
21.5 in.
13.8 in.
Polymer
21.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers