missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Adjustable Super Erecta™ Wire Shelves - Chrome Finish

For use with Metro™ Super Erecta™ adjustable shelving system

Brand:  Metro™ A2142NC

Product Code. 15956831

  • 127.00 € / Each
Estimated Shipment: 14-08-2025
to see stock.
Explore more special offers
Add to basket

Includes:

Each shelf shipped with one pack of four nonconductive sleeve/wedges.

This item is not returnable. View return policy

Description

Description

Each shelf provides approximately 76% open area for increased light penetration and improved visibility of stored items. Can be loaded and unloaded easily from all sides.

  • Hard, durable surface
  • Abrasion resistant
  • Ideal for dry, noncorrosive environments
  • Shelves adust in 1 in. increments with patented shelf-release corners
  • Shelf releases and wedges offer improved rigidity
  • Assembles quickly without tools; assembled units are nonconductive

Order Info

Posts sold separately.

Specifications

Specifications

42 x 21 in. (106.6 x 53 cm)
Metro™ Super Erecta™ adjustable shelving system
42 in.
21 in.
Chrome
Ideal for drying, non corrosive environments
Four non conductive sleeve/wedges
106.6 cm
53 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metro™ Super Adjustable Super Erecta™ Wire Shelves - Chrome Finish > 21 x 42 in. (53 x 107cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.