Learn More
Metro™ Super Adjustable Super Erecta™ Wire Shelf, Metroseal 3 Finish
Metroseal 3 wire shelf features a green epoxy finish with built-in Microban™ antimicrobial protection plus a unique open-wire design that permits free circulation of air and minimizes dust accumulation.
97.60 € - 223.00 €
Specifications
Finish | Metroseal Epoxy |
---|---|
For Use With (Equipment) | Metro™ Super Erecta™ adjustable shelving system |
Includes | Four non conductive sleeve/wedges |
Product Type | Wire Shelf |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15976841
|
Metro™
A2436NK3 |
36 x 24 in. (91.4 x 61 cm) |
136.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15956841
|
Metro™
A2430NK3 |
30 x 24 in. (76 x 61 cm) |
136.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15926851
|
Metro™
A2448NK3 |
48 x 24 in. (121.9 x 61 cm) |
163.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15926811
|
Metro™
A1830NK3 |
30 x 18 in. (76 x 45.7 cm) |
114.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15956821
|
Metro™
A1860NK3 |
60 x 18 in. (152.4 x 45.7 cm) |
156.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15906851
|
Metro™
A2442NK3 |
42 x 24 in. (106.6 x 61 cm) |
163.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15976851
|
Metro™
A2460NK3 |
60 x 24 in. (152.4 x 61 cm) |
192.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15906861
|
Metro™
A2472NK3 |
72 x 24 in. (182.9 x 61 cm) |
223.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15996811
|
Metro™
A1848NK3 |
48 x 18 in. (121.9 x 45.7 cm) |
135.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15996801
|
Metro™
A1824NK3 |
24 x 18 in. (61 x 45.7 cm) |
109.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15986821
|
Metro™
A1872NK3 |
72 x 18 in. (182.9 x 45.7 cm) |
179.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15946811
|
Metro™
A1836NK3 |
36 x 18 in. (91.4 x 45.7 cm) |
114.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15926821
|
Metro™
A1854NK3 |
54 x 18 in. (137 x 45.7 cm) |
156.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15976811
|
Metro™
A1842NK3 |
42 x 18 in. (106.6 x 45.7 cm) |
135.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15916801
|
Metro™
A1436NK3 |
36 x 14 in. (91.4 x 35.5 cm) |
102.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15986791
|
Metro™
A1430NK3 |
30 x 14 in. (76 x 35.5 cm) |
102.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15966791
|
Metro™
A1424NK3 |
24 x 14 in. (61 x 35.5 cm) |
97.85 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15976801
|
Metro™
A1460NK3 |
60 x 14 in. (152.4 x 35.5 cm) |
134.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15936801
|
Metro™
A1442NK3 |
42 x 14 in. (106.6 x 35.5 cm) |
118.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15956801
|
Metro™
A1448NK3 |
48 x 14 in. (121.9 x 35.5 cm) |
118.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15986801
|
Metro™
A1472NK3 |
72 x 14 in. (182.9 x 35.5 cm) |
152.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15926831
|
Metro™
A2130NK3 |
30 x 21 in. (76 x 53 cm) |
97.60 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15996831
|
Metro™
A2160NK3 |
60 x 21 in. (152.4 x 53 cm) |
176.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15956851
|
Metro™
A2454NK3 |
54 x 24 in. (182.9 x 61 cm) |
192.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
Description
- Each shelf provides approximately 76% open area.
- Can be loaded and unloaded easily from all sides.
- Corrosion-resistant carbon steel.
- Plated with self-sealing zinc and coated with an epoxy finish with Microban™ antimicrobial protection.
- Ideal for use in walk-in coolers and other environments that require corrosion resistance.
- Shelves adjust in 1 inch increments with patented shelf release corners.
- Shelf releases and wedges offer improved rigidity.
- Assembles quickly without tools; assembled units are nonconductive.
- Combine with Super Erecta Stationary or Mobile posts and accessories to create custom configurations for storage and transportation.
- Patented corner release system allows for easy height adjustment.
- Custom position your shelves for maximum storage density.
- Shelves up to 48 in. wide; 800 lbs per shelf capacity (evenly distributed).
- Shelves wider than 48 in.; 600 lbs per shelf (evenly distributed).
Order Info
Shelves and posts sold separately.
Warranty
12-year limited against rust.
Specifications
Metroseal Epoxy | |
Metro™ Super Erecta™ adjustable shelving system | |
Four non conductive sleeve/wedges | |
Wire Shelf |