Learn More
Metro™ Super Adjustable™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelf in stainless steel has a unique open-wire design permiting free circulation of air and minimizes dust accumulation.
203.00 € - 559.00 €
Specifications
For Use With (Equipment) | Metro™ Super Erecta™ adjustable shelving system |
---|---|
Includes | Four non conductive sleeve/wedges |
Product Type | Wire Shelf |
Material | Stainless Steel |
Requires: SiteSelect™ Posts (sold separately).
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15906811
|
Metro™
A1824NS |
24 x 18 in. (61 x 45.7 cm) |
245.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15936811
|
Metro™
A1830NS |
30 x 18 in. (76 x 45.7 cm) |
249.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15986811
|
Metro™
A1842NS |
42 x 18 in. (106.6 x 45.7 cm) |
295.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15956811
|
Metro™
A1836NS |
36 x 18 in. (91.4 x 45.7 cm) |
249.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15906821
|
Metro™
A1848NS |
48 x 18 in. (121.9 x 45.7 cm) |
295.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15966821
|
Metro™
A1860NS |
60 x 18 in. (152.4 x 45.7 cm) |
371.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15986841
|
Metro™
A2436NS |
36 x 24 in. (91.4 x 61 cm) |
307.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15916861
|
Metro™
A2472NS |
72 x 24 in. (182.9 x 61 cm) |
559.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15966851
|
Metro™
A2454NS |
54 x 24 in. (137 x 61 cm) |
447.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15936851
|
Metro™
A2448NS |
48 x 24 in. (121.9 x 61 cm) |
366.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15986851
|
Metro™
A2460NS |
60 x 24 in. (152.4 x 61 cm) |
447.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15936821
|
Metro™
A1854NS |
54 x 18 in. (137 x 45.7 cm) |
371.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15996821
|
Metro™
A1872NS |
72 x 18 in. (182.9 x 45.7 cm) |
438.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15916841
|
Metro™
A2172NS |
72 x 21 in. (182.9 x 53 cm) |
528.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15996791
|
Metro™
A1430NS |
30 x 14 in. (76.2 x 35.5 cm) |
210.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15976791
|
Metro™
A1424NS |
24 x 14 in. (61 x 35.5 cm) |
203.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15926801
|
Metro™
A1436NS |
36 x 14 in. (91.4 x 35.5 cm) |
210.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15916831
|
Metro™
A2124NS |
24 x 2 in. (61 x 53 cm) |
292.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15966801
|
Metro™
A1448NS |
48 x 14 in. (122 x 35.5 cm) |
254.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15946801
|
Metro™
A1442NS |
42 x 14 in. (106.6 x 35.5 cm) |
254.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15936831
|
Metro™
A2130NS |
30 x 2 in. (76.2 x 53 cm) |
295.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15976831
|
Metro™
A2154NS |
54 x 2 in. (137 x 53 cm) |
443.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15936841
|
Metro™
A2424NS |
24 x 24 in. (61 x 61 cm) |
302.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15916851
|
Metro™
A2442NS |
42 x 24 in. (106.6 x 61 cm) |
366.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15926861
|
Metro™
A3036NS |
36 x 30 in. (91.4 x 76.2 cm) | N/A | ||||||
15936861
|
Metro™
A3048NS |
48 x 30 in. (122 x 76.2 cm) | N/A | ||||||
15956861
|
Metro™
A3660NS |
60 x 36 in. (152.4 x 91.4 cm) | N/A | ||||||
15946861
|
Metro™
A3072NS |
72 x 30 in. (183 x 76.2 cm) | N/A | ||||||
15966861
|
Metro™
A3672NS |
72 x 36 in. (183 x 91.4 cm) | N/A | ||||||
Description
- Shelves adjust in 1 in. increments with patented shelf release corners
- Shelf releases and wedges offer improved rigidity
- Assembles quickly without tools
- Assembled units are nonconductive
- Maximum corrosion resistance
- Nonporous surface is easily cleaned
- Combine with Super Erecta Stationary or Mobile posts and accessories to create custom configurations for storage and transportation
- Patented corner release system allows for easy height adjustment
- Custom position your shelves for maximum storage density
- Capacity: Shelves up to 48 in. wide; 800 lbs per shelf capacity (evenly distributed)
- Shelves wider than 48 in.; 600 lbs per shelf (evenly distributed)
Services
One year
Specifications
Metro™ Super Erecta™ adjustable shelving system | |
Four non conductive sleeve/wedges | |
Wire Shelf | |
Stainless Steel |