Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Wire shelves are easily adjustable and the Metroseal 3 green epoxy with Microban™ finish promotes easy cleaning. For use with Metro Super Erecta shelving systems.
Brand:  Metro™ 1436NK3
 View more versions of this product
Product Code. 15946571
  • 89.20 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Corrosion resistant finish for wet or high humidity environments
  • Antimicrobial is built into finish to keep product clean
Specifications
Specifications
Wire Shelf
14 x 36 in.
Metroseal 3 with Microban