Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Brand:  Metro™ 2172NS
 View more versions of this product
Product Code. 15996641
  • 380.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
Wire Shelf
530 mm
Metro™ Super Erecta™ shelving system
Stainless Steel
21 in.
53 x 182.9 in. (21 x 72 cm)
Plastic sleeves and wedges