missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Wire Shelving Track Sets
For use with Metro Super Erecta™, Super Erecta Pro™ and Super Adjustable™ Top Track™ shelving.
Brand: Metro™ TTS20NA
This item is not returnable.
View return policy
Description
- Track sets include necessary sections of track for assembling track runs up to 21 ft. (640 cm)
- Coated with Microban™ antimicrobial protection
Order Info
19-138-228 (TTS2.5NA) includes a joining kit to connect the track to other track sets.
Specifications
Track Set | |
Necessary sections of track for assembling track runs of 1830 to 6405mm | |
609.6 cm |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ top track shelving | |
240 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Metro™ Wire Shelving Track Sets > Length: 609.6cm (20 ft.)
Spot an opportunity for improvement?Share a Content Correction