missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessory for Titrator: Titration Vessels

The right beaker for the right sample

Brand:  Mettler Toledo™ 00101446

 View more versions of this product

Product Code. 13126734

  • 1473.14 € / Pack of 20
Estimated Shipment: 02-10-2024
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

  • 09-313-151D: 250mL glass beakers for attachment directly to titration stand (set of 10)
  • 09-313-151Q: 80mL glass beakers (set of 20)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL Glass Titration Vessel, Pack of 20
Glass
Titrator
Titration
80 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers