Learn More
Mettler Toledo™ Adapters for pH Meters
Accessory adapters
Brand: Mettler Toledo™ 51302332
This item is not returnable.
View return policy
Description
LTW-MiniDin Conductivity Adapter
- The appropriate cable to connect the conductivity sensor mini DIN to a portable meter. Adapter is required for the following meters: FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro.
Infrared USB Adapter
- Infrared adapter to connect the SevenGo Duo to your computer over USB
Specifications
Infrared USB Adapter | |
Infrared adapter to connect the SevenGo Duo to your computer over USB | |
SevenGo Duo Pro |