missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Buffer Solutions: Standard pH
Storage and refill solutions for pH electrodes
Brand: Mettler Toledo™ 51350058
26.90 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
- Completely miscible
- Comes with Quality inspection certificate
- NIST traceable
Specifications
∼100°C | |
DIN | |
pH Buffer | |
250 mL Bottle | |
Completely Miscible |
Clear | |
Buffer Solution | |
Quality inspection certificate | |
10.012 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction