missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Cap Kits and Solutions for Portable Dissolved Oxygen Meters

For use with Portable Dissolved Oxygen Meters.

Brand:  Mettler Toledo™ 51300140


 View more versions of this product

Product Code. 12651316

This item is not returnable. View return policy

Description

Description

  • Zero Oxygen Standard
  • Capacity: 500mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 mL
Portable Dissolved Oxygen Meter
DO
DO zero oxygen tablets
Calibration Standard
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • zero oxygen tablets

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : adipic acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers