missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™
GREENER_CHOICE

Recommended NIST traceable solutions for calibrating or verifying METTLER TOLEDO conductivity electrodes.

79.05 € - 135.00 €

Specifications

Solution Type Conductivity Standard
Type Conductivity Standard
For Use With (Equipment) Conductivity Electrodes
View More Specs

Products 3
Product Code Brand Quantity Conductivity Price Quantity & Availability  
Product Code Brand Quantity Conductivity Price Quantity & Availability  
10528632
GREENER_CHOICE
View Documents
Mettler Toledo
51302050
20 mL 12.88 mS/cm
79.05 €
600mL
Please to purchase this item. Need a web account? Register with us today!
10166380
GREENER_CHOICE
View Documents
Mettler Toledo
51302049
20 mL 1413 μS/cm
79.35 €
600mL
Please to purchase this item. Need a web account? Register with us today!
15574843
View Documents
Mettler Toledo
30111140
20 mL 84 μS/cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • For daily verification: to monitor health of electrode and ensure measurement accuracy
  • For initial calibration of nominal cell constant electrodes
  • Be sure to screw cap tightly immediately after use (sachets minimize the chance of error due to environmental interaction)
  • Conductivity standards subject to change values when exposed to environment; water evaporation raises conductivity values
Specifications

Specifications

Conductivity Standard
Conductivity Standard
Conductivity Electrodes
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.