missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Replacement Tubing for DL31 Karl Fischer™ Titrators
Replacement parts for use with DL31 Karl Fischer titrators.
33.10 €
Specifications
Type | Solvent Delivery/Drain Tube with Screw Fitting |
---|---|
Includes | Solvent Dispensing Tubes, Suction Pipe |
For Use With (Equipment) | Mettler Toledo V20/V30 KF Titrators |
Description
For DL31 Karl Fischer titrators.Specifications
Solvent Delivery/Drain Tube with Screw Fitting | |
Solvent Dispensing Tubes, Suction Pipe | |
Mettler Toledo V20/V30 KF Titrators |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title