missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessories for FiveEasy Meters

For use with FiveEasy meters.

Brand:  Mettler Toledo™ 51302950

Product Code. 10493445

  • 84.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Provides 9VDC to meter
  • 100-240V 50/60Hz
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Power Adapter
pH
5 Connectors FiveEasy™
FiveEasy meters
100/240 V, 50/60 Hz
50/60 Hz
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers