missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Mono Half-Cell pH Measuring Electrodes
A half-cell pH measuring electrode featuring a glass body that is suited for high temperature and high pH samples in which the lifetime of the electrode is significantly shorter than that of a standard reference electrode.
172.00 € - 324.00 €
Specifications
Connector Types | S7 to BNC (30281915) |
---|---|
Material | Glass |
For Use With (Application) | Electrolyte hose coupling, Electrolyte bridge |
Length (English) | 4.7 in. |
Length (Metric) | 120 mm |
Product Code | Brand | Model | Type | Temperature (Metric) Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Temperature (Metric) Range | Price | Quantity & Availability | |||
10202903
|
METTLER TOLEDO™
51343195 |
InLab™ Mono | Half-Cell Measuring Electrode; HA Membrane Glass | 0°C to 100°C |
172.00 €
Each |
In Stock
Log in to see stock. |
|||
10496313
|
METTLER TOLEDO™
51343196 |
InLab™ Mono Plus | Half-Cell Measuring Electrode; A41 Thick-Walled Membrane Glass | 0°C to 130°C |
324.00 €
Each |
In Stock
Log in to see stock. |
|||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- High temperature samples
- High pH samples
- Corrosive acids and bases
- Drinking water
Accessories & Care
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 to BNC (30281915) | |
Electrolyte hose coupling, Electrolyte bridge | |
120 mm | |
12 mm |
Glass | |
4.7 in. | |
0.47 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title