missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Viscous pH Electrodes

A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.

533.00 € - 805.00 €

Specifications

Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 130°C
For Use With (Application) Highly viscous, highly glutinous samples
View More Specs

Products 3
Product Code Brand Model Type Connector Types For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Model Type Connector Types For Use With (Equipment) Price Quantity & Availability  
12570137
View Documents
METTLER TOLEDO™
51343150
InLab™ Viscous Combination Electrode S7 S7 to BNC (30281915)
533.00 €
Each
In Stock
to see stock.
Add to basket
 
10335303
View Documents
METTLER TOLEDO™
51343150
InLab™ Viscous Combination Electrode S7 S7 to BNC (30281915)
533.00 €
Each
In Stock
to see stock.
Add to basket
 
11977841
View Documents
METTLER TOLEDO™
51343151
InLab™ Viscous Pro-ISM Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology Multi-Pin Multi-Pin to BNC/RCA (30281896)
805.00 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Cosmetics
  • Gels, soaps, shampoos
  • Resins
  • Paints
  • Varnishes and glues
  • Colorants and dyes
  • Oily samples
  • TRIS compatible

Accessories & Care

  • Ceramic Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

Ceramic
0 to 14
Highly viscous, highly glutinous samples
HA Glass
32°F to 266°F
1.57 in.
Glass
0°C to 130°C
SteadyForce
3 mol/L KCl
0°C to 100°C
40 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.