missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Reference Electrodes
A reference electrode to be paired with half-cell measurement electrodes.
171.00 € - 348.00 €
Specifications
Connector Types | S7 |
---|---|
Material | Glass |
Filling Solution Type | 3mol/L KCl |
For Use With (Application) | For routine applications and measurements in Ion-selective half cells |
Length (English) | 4.72 in. |
Product Code | Brand | Model | Type | Temperature (Metric) Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Temperature (Metric) Range | Price | Quantity & Availability | |||
10018622
|
METTLER TOLEDO™
51343191 |
InLab™ Reference Plus | Half-Cell Reference Electrode | 0°C to 60°C |
327.00 €
Each |
In Stock
Log in to see stock. |
|||
10756513
|
METTLER TOLEDO™
51343190 |
InLab™ Reference | Half-Cell Reference Electrode | 0°C to 100°C |
171.00 €
Each |
In Stock
Log in to see stock. |
|||
10777304
|
METTLER TOLEDO™
51343192 |
InLab™ Reference Flow | Half-Cell Reference Electrode; Internal Gel Electrolyte | 0°C to 130°C |
348.00 €
Each |
In Stock
Log in to see stock. |
|||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- For use with half-cell measuring electrodes
Accessories & Care
- S7 connector type: Cable S7 to 2 mm Banana (30281923) required
- InLab Reference: ceramic junction
- InLab Reference Plus: moveable PTFE sleeve junction
- InLab Reference Flow: triple ceramic junction
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 | |
3mol/L KCl | |
4.72 in. | |
0.47 in. |
Glass | |
For routine applications and measurements in Ion-selective half cells | |
12 cm | |
1.2 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title