missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions

For use with ISE combination electrodes.

Brand:  Mettler Toledo™ 51340034

32.75 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12619195

  • 33.60 € / 20mL
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with ISE combination electrodes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Electrolyte for Nitrate ISE, 20mL
Ion-Selective Combination Electrodes Filling Solutions
Determine the Nitrate concentration in samples
32°F to 122°F
20 mL
ISE Combination Electrode
0.5 mol/L MgSO4
20 mL
0°C to 50°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.