missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ ISA Solutions
Solutions for perfectION™ combined ion selective electrodes
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11947511
|
Mettler Toledo™
51344760 |
perfectION ISA solid state ISE | 475 mL |
N/A
|
N/A | ||||
11957511
|
Mettler Toledo™
51344761 |
perfectION calcium ISA | 475 mL |
N/A
|
N/A | ||||
11977511
|
Mettler Toledo™
51344763 |
perfectION nitrate ISA | 475 mL |
N/A
|
N/A | ||||
12797421
|
Mettler Toledo™
51344762 |
perfectION potassium ISA | 475 mL |
N/A
|
N/A | ||||
11987511
|
Mettler Toledo™
51344764 |
perfectION nitrate ISS | 475 mL |
N/A
|
N/A | ||||
11997511
|
Mettler Toledo™
51344765 |
perfectION TISAB II with CDTA | 3790 mL |
N/A
|
N/A | ||||
11907521
|
Mettler Toledo™
51344766 |
perfectION TISAB III with CDTA | 475 mL |
N/A
|
N/A | ||||
Description
Solutions (ionic-strength adjustment buffers)
- Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
- ISA for calcium electrode, 475mL
- ISA for potassium electrode, 475mL
- ISA for nitrate electrode, 475mL
- Nitrate ISS (for suppressing interference), 475mL
- Fluoride TISAB II with CDTA, 3790mL
- Fluoride TISAB III with CDTA (concentrate), 475mL
Specifications
ISA Solution |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.