missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH Electrode Accessories: Glass Flow Cell

For use with electrode with 12mm dia. shaft

Brand:  Mettler Toledo™ 51302257

Product Code. 10750513

This item is not returnable. View return policy

Description

Description

For more accurate measurements in ultra pure water
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glass flow-through cell
Electrodes with 12 mm dia. Shaft
Flow Cell
Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers