Learn More
METTLER TOLEDO™ SevenExcellence™ S400 Benchtop pH Meter
The S400 meter measures pH, redox/ORP, and ISFET. It can be expanded with additional modules to measure conductivity, resistivity, DO, BOD, ISE, salinity, and TDS. It has an intuitive touchscreen, the ability to create measurement shortcuts, and can export data to a printer, Excel, or LIMS system.
Brand:  METTLER TOLEDO™ 30046242
2291.25 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 13420679
  • 3260.00 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • pH, Redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, ISE, salinity, TDS, and BOD
  • Easily create measurement and calibration shortcuts
  • 7 inch color touch screen with intuitive menu navigation
  • Compatible with a wide range of electrodes for any application
  • *Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Export data to a printer, Excel file, or LIMS system
  • Automation available for measurements and cleaning in high-throughput applications
  • Manage user permissions and access to ensure data integrity
  • Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
  • Optional comprehensive qualification package available including IQ/OQ/PQ
  • Preventative maintenance and service available through METTLER TOLEDO Service
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S400 pH meter; InLab Routine Pro-ISM electrode; electrode holder; ; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate; pH measurement guide; Multipin-BNC/RCA electrode cable 1.2 m
±0.1
USB, RS-232, Ethernet
0.001, 0.01, 0.1
-2000 to 2000
-30°C to +130°C
TFT
20,000
0.001μS/cm to 2000mS/cm
0.001 to 1
13 pre-defined and 20 user-defined standard
±0.1°C
±0.002
-2 to 20
Max. 5 points, 8 pre-defined and 20 user-defined buffer group
0.1/1mV
0.1°C
250 analyses (20.000 data points)
S400 Kit - Bio & Life Science Applications
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Single-channel